siRNA Details
Gene Symbol: LOC494141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026541.1 | 2 | 1147 | |||
NR_026563.1 | 1 | 1292 | ||||
NR_026564.1 | 2 | 1147 | ||||
GenBank mRNA | AK131059.1 | - | 2 | 1125 | ||
BC026079.1 | - | 1 | 1299 | |||
BC026082.1 | - | 2 | 1144 | |||
BC034967.1 | - | 1 | 1299 | |||
BC044592.1 | - | 1 | 1292 | |||
BC056262.1 | - | 2 | 1149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
