siRNA Details
Gene Symbol: C5orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144954.1 | NP_001138426.1 | 2 | 481 | ||
XM_011534431.1 | XP_011532733.1 | 2 | 475 | |||
XM_017009028.1 | XP_016864517.1 | 2 | 475 | |||
GenBank mRNA | BC034971.1 | - | 2 | 447 | ||
BC047225.1 | - | 2 | 436 | |||
BC053953.1 | - | 2 | 436 | |||
BC057842.1 | - | 2 | 422 | |||
BC066964.1 | - | 2 | 443 | |||
BC092489.1 | - | 2 | 470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
