siRNA Details
Gene Symbol: C5orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144954.1 | NP_001138426.1 | 3, 4 | 619 | ||
XM_017009028.1 | XP_016864517.1 | 3, 4 | 613 | |||
GenBank mRNA | BC034971.1 | - | 3, 4 | 585 | ||
BC047225.1 | - | 3, 4 | 574 | |||
BC053953.1 | - | 3, 4 | 574 | |||
BC057842.1 | - | 3, 4 | 560 | |||
BC066964.1 | - | 3, 4 | 581 | |||
BC092489.1 | - | 3, 4 | 608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
