siRNA Details
Gene Symbol: CCDC169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144981.2 | NP_001138453.1 | 2 | 255 | ||
NM_001198908.1 | NP_001185837.1 | 2 | 255 | |||
GenBank mRNA | AK131058.1 | - | 2 | 206 | ||
BC146975.1 | - | 2 | 149 | 9056903 | ||
DB454585.1 | - | 2 | 256 | |||
DN999189.1 | - | 2 | 212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
