siRNA Details
Gene Symbol: FAM26F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010919.2 | NP_001010919.1 | 3 | 706 | ||
NM_001276460.1 | NP_001263389.1 | 2 | 123 | |||
XM_005266999.3 | XP_005267056.1 | 2 | 990 | |||
XM_011535845.2 | XP_011534147.1 | 2 | 1070 | |||
GenBank mRNA | AF086130.1 | - | 1 | 28 | ||
AV734646.2 | - | 2 | 123 | |||
BC146842.1 | - | 2 | 632 | 9056770 | ||
DN997293.1 | - | 2 | 622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
