siRNA Details
Gene Symbol: WHAMMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026589.1 | 2 | 680 | |||
GenBank mRNA | BC035099.1 | - | 2 | 247 | ||
BC066982.1 | - | 2 | 421 | |||
BC150524.1 | - | 1 | 86 |
Gene Symbol: WHAMMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036650.1 | 1 | 450 | |||
GenBank mRNA | BC117556.1 | - | 450 |
Gene Symbol: WHAMMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003521.1 | 2 | 673 | |||
GenBank mRNA | AL833132.1 | - | 2 | 284 | ||
BC018097.1 | - | 2 | 151 | |||
BC044610.1 | - | 2 | 284 | |||
BC048987.1 | - | 2 | 733 | |||
DQ309039.1 | - | 2 | 141 | |||
DQ309040.1 | - | 2 | 673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
