siRNA Details
Gene Symbol: RGS7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029875.2 | NP_001025046.1 | 5 | 1301 | ||
NM_001271890.1 | NP_001258819.1 | 3 | 415 | |||
NM_001271891.1 | NP_001258820.1 | 2, 3 | 413 | |||
NR_073511.1 | 3 | 415 | ||||
XM_005248502.3 | XP_005248559.1 | 4, 5 | 2681 | |||
GenBank mRNA | AK294808.1 | - | 4 | 406 | ||
AK296443.1 | - | 5 | 949 | |||
AK296494.1 | - | 3 | 383 | |||
BC140707.1 | - | 5 | 647 | 9021575 | ||
BX640900.1 | - | 5 | 539 | |||
DA211241.1 | - | 3 | 416 | |||
DA417118.1 | - | 3 | 383 | |||
DC342795.1 | - | 2, 3 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
