siRNA Details
Gene Symbol: RGS7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029875.2 | NP_001025046.1 | 2 | 860 | ||
XM_005248502.3 | XP_005248559.1 | 2 | 2242 | |||
GenBank mRNA | AK296443.1 | - | 2 | 508 | ||
BC140707.1 | - | 2 | 206 | 9021575 | ||
BX640900.1 | - | 2 | 98 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
