siRNA Details
Gene Symbol: KRTAP24-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085455.2 | NP_001078924.1 | 1 | 595 | ||
GenBank mRNA | AB096935.1 | - | 1 | 595 | ||
AM238513.1 | - | 1 | 569 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
