siRNA Details
Gene Symbol: OR6W1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002140.1 | 1 | 760 | |||
GenBank mRNA | BC069382.1 | - | 1 | 351 | ||
BC074961.2 | - | 1 | 336 | |||
BC074962.2 | - | 1 | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
