siRNA Details
Gene Symbol: TMEM201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010866.3 | NP_001010866.1 | 1, 2 | 181 | ||
NM_001130924.2 | NP_001124396.2 | 1, 2 | 181 | |||
XM_006710417.3 | XP_006710480.1 | 1, 2 | 212 | |||
XM_011540910.2 | XP_011539212.1 | 1, 2 | 210 | |||
XM_017000549.1 | XP_016856038.1 | 1, 2 | 212 | |||
XM_017000550.1 | XP_016856039.1 | 1, 2 | 212 | |||
XM_017000551.1 | XP_016856040.1 | 1, 2 | 422 | |||
XM_017000552.1 | XP_016856041.1 | 1, 2 | 250 | |||
GenBank mRNA | BC121187.1 | - | 1, 2 | 136 | 3870153 | |
BC137291.1 | - | 1, 2 | 181 | 9021288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
