siRNA Details
Gene Symbol: SPANXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009615.2 | NP_001009615.1 | 1 | 17 | ||
GenBank mRNA | DQ336112.1 | - | 1 | 17 | ||
DQ336113.1 | - | 1 | 17 | |||
DQ336114.1 | - | 1 | 17 | |||
DQ336115.1 | - | 1 | 17 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
