siRNA Details
Gene Symbol: PTAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099666.1 | NP_001093136.1 | 5 | 630 | ||
XM_005251976.4 | XP_005252033.1 | 5 | 720 | |||
XM_005251977.4 | XP_005252034.1 | 5 | 694 | |||
XM_005251980.4 | XP_005252037.1 | 5 | 680 | |||
XM_005251981.4 | XP_005252038.1 | 5 | 680 | |||
XM_011518639.1 | XP_011516941.1 | 5 | 594 | |||
XM_011518640.1 | XP_011516942.1 | 6 | 770 | |||
GenBank mRNA | AL832683.1 | - | 3 | 374 | ||
BC053622.1 | - | 5 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
