siRNA Details
Gene Symbol: PTAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099666.1 | NP_001093136.1 | 8 | 1140 | ||
XM_005251976.4 | XP_005252033.1 | 7 | 1311 | |||
XM_005251977.4 | XP_005252034.1 | 8 | 1201 | |||
XM_005251980.4 | XP_005252037.1 | 7 | 885 | |||
XM_005251981.4 | XP_005252038.1 | 7 | 882 | |||
XM_011518639.1 | XP_011516941.1 | 7 | 1185 | |||
XM_011518640.1 | XP_011516942.1 | 8 | 1361 | |||
GenBank mRNA | AK127511.1 | - | 2 | 475 | ||
AL832683.1 | - | 5 | 965 | |||
BC053622.1 | - | 8 | 1122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
