siRNA Details
Gene Symbol: NBPF13P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103466.1 | 4 | 458 | |||
GenBank mRNA | BX336604.2 | - | 4 | 458 | ||
CB154168.1 | - | 2 | 135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
