siRNA Details
Gene Symbol: CEACAM18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278392.1 | NP_001265321.1 | 3 | 605 | ||
XM_011527258.2 | XP_011525560.1 | 3 | 705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
