siRNA Details
Gene Symbol: RRP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015056.2 | NP_055871.1 | 5, 6 | 541 | ||
GenBank mRNA | AK056263.1 | - | 5, 6 | 504 | ||
BC014005.2 | - | 5, 6 | 513 | |||
BC028386.2 | - | 5, 6 | 546 | 4821672 | ||
D80001.1 | - | 5, 6 | 439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
