siRNA Details
Gene Symbol: ARHGEF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242729.1 | NP_001229658.1 | 13 | 2121 | ||
XM_006714244.3 | XP_006714307.1 | 11 | 1481 | |||
XM_011532050.2 | XP_011530352.1 | 12 | 1670 | |||
GenBank mRNA | AK125951.1 | - | 4 | 599 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
