siRNA Details
Gene Symbol: MAGEB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099921.1 | NP_001093391.1 | 2 | 620 | ||
XM_011545453.2 | XP_011543755.1 | 2 | 509 | |||
XM_011545454.2 | XP_011543756.1 | 2 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
