siRNA Details
Gene Symbol: FAM205C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001309426.1 | NP_001296355.1 | 5 | 878 | ||
NM_001309427.1 | NP_001296356.1 | 5 | 488 | |||
XM_017014165.1 | XP_016869654.1 | 5 | 491 | |||
XM_017014166.1 | XP_016869655.1 | 5 | 427 | |||
GenBank mRNA | DB042441.1 | - | 5 | 490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
