siRNA Details
Gene Symbol: PALM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145028.1 | NP_001138500.1 | 6 | 415 | ||
XM_005259891.4 | XP_005259948.1 | 7 | 616 | |||
XM_011527965.2 | XP_011526267.1 | 6 | 536 | |||
GenBank mRNA | AW130597.1 | - | 4 | 393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
