siRNA Details
Gene Symbol: TNRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080495.2 | NP_001073964.2 | 30 | 9067 | ||
XM_017012728.1 | XP_016868217.1 | 31 | 11597 | |||
XM_017012729.1 | XP_016868218.1 | 30 | 9304 | |||
XM_017012730.1 | XP_016868219.1 | 30 | 8919 | |||
XM_017012731.1 | XP_016868220.1 | 29 | 8794 | |||
XM_017012732.1 | XP_016868221.1 | 29 | 8791 | |||
XM_017012733.1 | XP_016868222.1 | 29 | 8779 | |||
XM_017012734.1 | XP_016868223.1 | 30 | 9805 | |||
XM_017012735.1 | XP_016868224.1 | 30 | 8807 | |||
XM_017012736.1 | XP_016868225.1 | 30 | 9394 | |||
XM_017012737.1 | XP_016868226.1 | 30 | 11167 | |||
XM_017012738.1 | XP_016868227.1 | 30 | 8689 | |||
GenBank mRNA | AB058759.1 | - | 14 | 3722 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
