siRNA Details
Gene Symbol: MAPKAPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020127.1 | XP_016875616.1 | 15 | 8067 | ||
XM_017020128.1 | XP_016875617.1 | 15 | 6518 | |||
XM_017020132.1 | XP_016875621.1 | 16 | 8115 | |||
XM_017020133.1 | XP_016875622.1 | 15 | 7887 | |||
XM_017020134.1 | XP_016875623.1 | 15 | 7713 | |||
XM_017020135.1 | XP_016875624.1 | 14 | 7796 | |||
XM_017020136.1 | XP_016875625.1 | 15 | 7881 | |||
XM_017020138.1 | XP_016875627.1 | 11 | 7274 |
Gene Symbol: ADAM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036636.1 | 1 | 1872 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
