siRNA Details
Gene Symbol: ZNF780A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010880.2 | NP_001010880.2 | 6 | 2779 | ||
NM_001142577.1 | NP_001136049.1 | 6 | 2705 | |||
NM_001142578.1 | NP_001136050.1 | 6 | 2702 | |||
XM_005258773.2 | XP_005258830.1 | 6 | 2782 | |||
XM_006723150.3 | XP_006723213.1 | 5 | 3402 | |||
XM_011526771.2 | XP_011525073.1 | 6 | 2824 | |||
XM_011526772.2 | XP_011525074.1 | 6 | 2836 | |||
XM_017026611.1 | XP_016882100.1 | 6 | 3489 | |||
XM_017026612.1 | XP_016882101.1 | 7 | 3628 | |||
XM_017026613.1 | XP_016882102.1 | 7 | 2991 | |||
XM_017026614.1 | XP_016882103.1 | 7 | 2979 | |||
XM_017026615.1 | XP_016882104.1 | 7 | 2933 | |||
XM_017026616.1 | XP_016882105.1 | 7 | 2857 | |||
XM_017026617.1 | XP_016882106.1 | 5 | 3331 | |||
XM_017026618.1 | XP_016882107.1 | 6 | 2829 | |||
XM_017026619.1 | XP_016882108.1 | 5 | 3395 | |||
GenBank mRNA | AK091274.1 | - | 6 | 2764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
