siRNA Details
Gene Symbol: HYPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012274.1 | NP_036406.1 | 1 | 264 | ||
GenBank mRNA | AF049615.1 | - | 1 | 264 | ||
BC104428.1 | - | 1 | 241 | 40021552 | ||
BC104429.1 | - | 1 | 241 | 40021554 | ||
BC113024.1 | - | 1 | 241 | 40021553 | ||
BC113025.1 | - | 1 | 241 | 40021555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
