siRNA Details
Gene Symbol: TMEM221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190844.1 | NP_001177773.1 | 2 | 440 | ||
XM_011527603.2 | XP_011525905.1 | 2 | 712 | |||
GenBank mRNA | BC039402.1 | - | 2 | 182 | ||
BY797043.2 | - | 2 | 440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
