siRNA Details
Gene Symbol: TMEM221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190844.1 | NP_001177773.1 | 3 | 681 | ||
XM_011527603.2 | XP_011525905.1 | 3 | 953 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
