siRNA Details
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 2, 3 | 875 | ||
XM_011536071.1 | XP_011534373.1 | 3, 4 | 657 | |||
GenBank mRNA | AK055077.1 | - | 2, 3 | 875 | ||
BX537882.1 | - | 2 | 446 | |||
EU518415.1 | - | 2, 3 | 277 | |||
FM205709.1 | - | 2, 3 | 724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
