siRNA Details
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 2 | 679 | ||
XM_011536071.1 | XP_011534373.1 | 3 | 461 | |||
GenBank mRNA | AK055077.1 | - | 2 | 679 | ||
BX537882.1 | - | 2 | 250 | |||
EU518415.1 | - | 2 | 81 | |||
FM205709.1 | - | 2 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
