siRNA Details
Gene Symbol: ZNF823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080493.3 | NP_001073962.1 | 4 | 2419 | ||
NM_001297610.1 | NP_001284539.1 | 3 | 2358 | |||
NM_017507.1 | NP_059977.1 | 3 | 2292 | |||
XM_017026941.1 | XP_016882430.1 | 4 | 2581 | |||
XM_017026942.1 | XP_016882431.1 | 2 | 2219 | |||
GenBank mRNA | AK303213.1 | - | 3 | 2323 | ||
BC052597.1 | - | 4 | 2331 | 4304302 | ||
BC063560.1 | - | 1 | 1592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
