siRNA Details
Gene Symbol: SH2D5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103160.1 | NP_001096630.1 | 9 | 3483 | ||
NM_001103161.1 | NP_001096631.1 | 10 | 3612 | |||
XM_011541459.1 | XP_011539761.1 | 10 | 4077 | |||
XM_011541460.1 | XP_011539762.1 | 11 | 3917 | |||
XM_011541461.2 | XP_011539763.1 | 10 | 3190 | |||
XM_011541462.1 | XP_011539764.1 | 10 | 4074 | |||
GenBank mRNA | AK123236.1 | - | 1 | 2028 | ||
AK123841.1 | - | 6 | 2841 | |||
AK124869.1 | - | 9 | 3483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
