siRNA Details
Gene Symbol: ENO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242699.1 | NP_001229628.1 | 3, 4 | 537 | ||
XM_005269815.3 | XP_005269872.1 | 3, 4 | 1819 | |||
XM_006717835.3 | XP_006717898.1 | 3, 4 | 1819 | |||
XM_006717836.3 | XP_006717899.1 | 2, 3 | 354 | |||
XM_011539797.2 | XP_011538099.1 | 3, 4 | 1819 | |||
XM_011539799.2 | XP_011538101.1 | 3, 4 | 1819 | |||
XM_017016241.1 | XP_016871730.1 | 3, 4 | 1819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
