siRNA Details
Gene Symbol: C2orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099334.2 | NP_001092804.1 | 3 | 277 | ||
XM_017004074.1 | XP_016859563.1 | 3 | 290 | |||
XM_017004075.1 | XP_016859564.1 | 3 | 225 | |||
XM_017004076.1 | XP_016859565.1 | 3 | 265 | |||
XM_017004077.1 | XP_016859566.1 | 3 | 225 | |||
XM_017004078.1 | XP_016859567.1 | 3 | 225 | |||
GenBank mRNA | BC035737.1 | - | 3 | 277 | 5730487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
