siRNA Details
Gene Symbol: SZRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114600.2 | NP_001108072.1 | 4 | 675 | ||
NM_001271869.1 | NP_001258798.1 | 4 | 672 | |||
NR_073500.1 | 2 | 370 | ||||
NR_073501.1 | 5 | 746 | ||||
NR_073502.1 | 3 | 625 | ||||
NR_073503.1 | 3 | 622 | ||||
GenBank mRNA | AB097042.1 | - | 3 | 413 | ||
AB097043.1 | - | 3 | 413 | |||
AB097044.1 | - | 3 | 404 | |||
AB097045.1 | - | 3 | 404 | |||
AB097046.1 | - | 4 | 463 | |||
AJ295987.1 | - | 3 | 409 | |||
AK308787.1 | - | 2 | 157 | |||
AL050028.1 | - | 3 | 409 | |||
BC023988.1 | - | 3 | 430 | 3825796 | ||
BC065538.1 | - | 3 | 410 | 6053254 | ||
DA010145.1 | - | 4 | 467 | |||
DA438597.1 | - | 5 | 533 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001025608, NM_001277195, BC066992, AK027934, AK171420, and others.
