siRNA Details
Gene Symbol: PERM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291366.1 | NP_001278295.1 | 4 | 3003 | ||
NM_001291367.1 | NP_001278296.1 | 5 | 2650 | |||
XM_017002583.1 | XP_016858072.1 | 4 | 3105 | |||
XM_017002584.1 | XP_016858073.1 | 4 | 3128 | |||
XM_017002585.1 | XP_016858074.1 | 3 | 3494 | |||
GenBank mRNA | AK123855.1 | - | 5 | 2626 | ||
BC006300.2 | - | 2 | 800 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
