siRNA Details
Gene Symbol: NANOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029861.2 | NP_001025032.1 | 1 | 114 | ||
GenBank mRNA | BC042883.1 | - | 1 | 116 | 5270224 | |
BC117484.1 | - | 1 | 206 | 40126035 | ||
BC117486.1 | - | 1 | 206 | 40126037 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
