siRNA Details
Gene Symbol: AK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145128.2 | NP_001138600.2 | 20 | 2223 | ||
XM_006715376.3 | XP_006715439.2 | 21 | 3088 | |||
XM_011535550.2 | XP_011533852.1 | 21 | 3088 | |||
XM_011535552.2 | XP_011533854.1 | 20 | 2264 | |||
XM_011535553.2 | XP_011533855.1 | 20 | 2871 | |||
XM_011535554.2 | XP_011533856.1 | 19 | 2767 | |||
XM_011535555.2 | XP_011533857.1 | 21 | 3088 | |||
XM_011535556.2 | XP_011533858.1 | 21 | 3088 | |||
XM_011535558.1 | XP_011533860.1 | 2 | 109 | |||
XM_011535559.2 | XP_011533861.1 | 21 | 3088 | |||
XM_017010384.1 | XP_016865873.1 | 21 | 3088 | |||
XM_017010385.1 | XP_016865874.1 | 20 | 2256 | |||
XM_017010386.1 | XP_016865875.1 | 21 | 3088 | |||
XM_017010387.1 | XP_016865876.1 | 15 | 2719 | |||
XM_017010388.1 | XP_016865877.1 | 21 | 3088 | |||
XM_017010389.1 | XP_016865878.1 | 6 | 971 | |||
GenBank mRNA | AK124171.1 | - | 8 | 857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
