siRNA Details
Gene Symbol: FAM196A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039762.2 | NP_001034851.1 | 6 | 2144 | ||
XM_005252694.3 | XP_005252751.1 | 4 | 2119 | |||
XM_017016537.1 | XP_016872026.1 | 4 | 5715 | |||
XM_017016538.1 | XP_016872027.1 | 5 | 5131 | |||
XM_017016539.1 | XP_016872028.1 | 4 | 2858 | |||
XM_017016540.1 | XP_016872029.1 | 5 | 2984 | |||
XM_017016541.1 | XP_016872030.1 | 4 | 2420 | |||
XM_017016542.1 | XP_016872031.1 | 5 | 5239 | |||
GenBank mRNA | AK127465.1 | - | 6 | 2144 | ||
BC137112.1 | - | 4 | 1726 | 9021109 | ||
BC144478.1 | - | 3 | 1654 | 9053005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
