siRNA Details
Gene Symbol: LBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009812.1 | NP_001009812.1 | 2 | 1034 | ||
NM_001282430.1 | NP_001269359.1 | 2 | 610 | |||
GenBank mRNA | AY203953.1 | - | 2 | 769 | ||
AY305861.1 | - | 2 | 1034 | |||
BC144442.1 | - | 3 | 883 | |||
BC150517.1 | - | 2 | 594 | 40132747 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
