siRNA Details
Gene Symbol: MYO1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101421.3 | NP_001094891.3 | 3 | 279 | ||
XM_011538223.2 | XP_011536525.1 | 3 | 721 | |||
XM_017019207.1 | XP_016874696.1 | 3 | 721 | |||
GenBank mRNA | DV080477.1 | - | 2 | 80 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
