siRNA Details
Gene Symbol: SKOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258024.1 | NP_001244953.1 | 10 | 2168 | ||
XM_017022173.1 | XP_016877662.1 | 4 | 2581 | |||
GenBank mRNA | AY669507.1 | - | 10 | 2168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
