siRNA Details
Gene Symbol: CCDC66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012506.4 | NP_001012524.4 | 9 | 1225 | ||
NM_001141947.1 | NP_001135419.1 | 9 | 1244 | |||
NR_024460.1 | 9 | 1225 | ||||
XM_005265081.3 | XP_005265138.1 | 9 | 1244 | |||
XM_005265082.3 | XP_005265139.1 | 9 | 1223 | |||
XM_005265084.3 | XP_005265141.1 | 9 | 1238 | |||
XM_011533614.2 | XP_011531916.1 | 8 | 1249 | |||
XM_011533615.2 | XP_011531917.1 | 8 | 1259 | |||
XM_011533616.2 | XP_011531918.1 | 9 | 1183 | |||
XM_011533617.2 | XP_011531919.1 | 10 | 1341 | |||
XM_011533619.2 | XP_011531921.1 | 10 | 1244 | |||
XM_017006229.1 | XP_016861718.1 | 9 | 1223 | |||
XM_017006230.1 | XP_016861719.1 | 8 | 1227 | |||
XM_017006231.1 | XP_016861720.1 | 8 | 1248 | |||
XM_017006234.1 | XP_016861723.1 | 9 | 1283 | |||
XM_017006235.1 | XP_016861724.1 | 10 | 1341 | |||
XM_017006236.1 | XP_016861725.1 | 11 | 1336 | |||
XM_017006237.1 | XP_016861726.1 | 11 | 1373 | |||
XM_017006238.1 | XP_016861727.1 | 12 | 1514 | |||
XM_017006239.1 | XP_016861728.1 | 6 | 574 | |||
XM_017006240.1 | XP_016861729.1 | 10 | 1295 | |||
XM_017006243.1 | XP_016861732.1 | 9 | 1210 | |||
XM_017006244.1 | XP_016861733.1 | 10 | 1346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
