siRNA Details
Gene Symbol: SYDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032184.1 | NP_115560.1 | 4, 5 | 2728 | ||
XM_005271254.3 | XP_005271311.1 | 3, 4 | 3279 | |||
XM_017002483.1 | XP_016857972.1 | 4, 5 | 3408 | |||
XM_017002484.1 | XP_016857973.1 | 4, 5 | 3407 | |||
XM_017002485.1 | XP_016857974.1 | 4, 5 | 2115 | |||
XM_017002486.1 | XP_016857975.1 | 4, 5 | 2028 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
