siRNA Details
Gene Symbol: SYDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032184.1 | NP_115560.1 | 7 | 3283 | ||
XM_017002483.1 | XP_016857972.1 | 7 | 3958 | |||
XM_017002485.1 | XP_016857974.1 | 7 | 2670 | |||
XM_017002486.1 | XP_016857975.1 | 7 | 2583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
