siRNA Details
Gene Symbol: SYDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032184.1 | NP_115560.1 | 6 | 2972 | ||
XM_017002483.1 | XP_016857972.1 | 6 | 3652 | |||
XM_017002484.1 | XP_016857973.1 | 6 | 3651 | |||
XM_017002485.1 | XP_016857974.1 | 6 | 2359 | |||
XM_017002486.1 | XP_016857975.1 | 6 | 2272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
