siRNA Details
Gene Symbol: NUGGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010906.1 | NP_001010906.1 | 19 | 3340 | ||
XM_011544523.2 | XP_011542825.1 | 19 | 3609 | |||
XM_011544524.2 | XP_011542826.1 | 19 | 5115 | |||
XM_011544525.1 | XP_011542827.1 | 11 | 2171 | |||
GenBank mRNA | AK129923.1 | - | 5 | 1441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
