siRNA Details
Gene Symbol: BCL2L15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010922.2 | NP_001010922.1 | 2, 3 | 416 | ||
GenBank mRNA | AI821052.1 | - | 2, 3 | 416 | ||
AK289439.1 | - | 2, 3 | 450 | |||
AY265861.1 | - | 2, 3 | 244 | |||
BC127719.1 | - | 2, 3 | 277 | 40127248 | ||
BC127720.1 | - | 2, 3 | 277 | 40127249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
