siRNA Details
Gene Symbol: TMEM202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080462.1 | NP_001073931.1 | 4 | 566 | ||
XM_011521497.1 | XP_011519799.1 | 4 | 567 | |||
XM_011521499.1 | XP_011519801.1 | 4 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
