siRNA Details
Gene Symbol: CENPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012267.2 | NP_001012267.1 | 8 | 2067 | ||
NM_001286969.1 | NP_001273898.1 | 7 | 1885 | |||
NM_001286971.1 | NP_001273900.1 | 4 | 1224 | |||
GenBank mRNA | AK091247.1 | - | 4 | 1225 | ||
AK092603.1 | - | 8 | 1882 | |||
AL833701.1 | - | 3 | 1742 | |||
BX537851.1 | - | 7 | 1413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
