siRNA Details
Gene Symbol: CENPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012267.2 | NP_001012267.1 | 3 | 850 | ||
NM_001286969.1 | NP_001273898.1 | 2 | 668 | |||
XM_011518685.2 | XP_011516987.1 | 3 | 861 | |||
XM_011518689.1 | XP_011516991.1 | 3 | 861 | |||
XM_017014715.1 | XP_016870204.1 | 3 | 861 | |||
GenBank mRNA | AK092603.1 | - | 3 | 665 | ||
AK307435.1 | - | 2 | 188 | |||
BC071726.1 | - | 3 | 376 | 5554572 | ||
BX537851.1 | - | 2 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
